Please login to your online account to display your discounted pricing

Propyltriphenylphosphonium iodide, 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material Propyltriphenylphosphonium iodide
Assay Percent Range 99%
Molecular Formula C21H22IP
Formula Weight 432.28
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC453040050

  SDS
thermo fisher scientific
AC453040050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC453040250

  SDS
thermo fisher scientific
AC453040250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Propyltriphenylphosphonium iodide
Quantity 5g
CAS 14350-50-6
Assay Percent Range 99%
Molecular Formula C21H22IP
Formula Weight 432.28