missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propyltriphenylphosphonium iodide, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC453040250

 View more versions of this product

Catalog No. AC453040250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Propyltriphenylphosphonium iodide
25g
99%
n-propyl-triphenyl phosphonium iodide, phosphonium,triphenylpropyl-, iodide 1:1, propyl triphenyl phosphonium iodide, propyltriphenylphosphonium iodide, triphenyl propyl phosphanium iodide, triphenyl propyl phosphonium iodide
CCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[I-]
432.28
432.28
206.0°C to 209.0°C
14350-50-6
C21H22IP
MPNQDJZRGAOBPW-UHFFFAOYSA-M
triphenyl(propyl)phosphanium;iodide
10982905
99%

Chemical Identifiers

14350-50-6
432.28
n-propyl-triphenyl phosphonium iodide, phosphonium,triphenylpropyl-, iodide 1:1, propyl triphenyl phosphonium iodide, propyltriphenylphosphonium iodide, triphenyl propyl phosphanium iodide, triphenyl propyl phosphonium iodide
triphenyl(propyl)phosphanium;iodide
C21H22IP
MPNQDJZRGAOBPW-UHFFFAOYSA-M
10982905
CCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[I-]
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.

GHS Signal Word: Warning

SDS Documents