Please login to your online account to display your discounted pricing

Propylmagnesium chloride, 2M solution in diethyleether, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC208451000

 View more versions of this product

Catalog No. AC208451000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 102.85
Quantity 100mL
Color Gray to red to brown
Packaging AcroSeal™ Glass Bottle
Density 0.8270g/mL
Chemical Name or Material Propylmagnesium chloride
Molecular Formula C3H7ClMg
Additional Information product may develop some turbidity
CAS 2234-82-4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Extremely flammable liquid and vapour.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.
Harmful if swallowed.
May cause drowsiness or dizziness.
Reacts violently with water.
May form explosive peroxides.
Repeated ex
CAS 60-29-7
Linear Formula CH3CH2CH2MgCl
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Propylmagnesium chloride
CAS 2234-82-4
CAS 60-29-7
Molecular Formula C3H7ClMg
Linear Formula CH3CH2CH2MgCl
Formula Weight 102.85
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Extremely flammable liquid and vapour.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.
Harmful if swallowed.
May cause drowsiness or dizziness.
Reacts violently with water.
May form explosive peroxides.
Repeated ex