Please login to your online account to display your discounted pricing

Propylmagnesium chloride, 2M solution in diethyleether, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Propylmagnesium chloride
CAS 2234-82-4
CAS 60-29-7
Molecular Formula C3H7ClMg
Linear Formula CH3CH2CH2MgCl
Formula Weight 102.85g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC208458000

 
thermo fisher scientific
208458000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC208451000

 
thermo fisher scientific
208451000
AcroSeal™ Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Additional Information product may develop some turbidity
Packaging AcroSeal™ Glass Bottle
Density 0.8270g/mL
Color Gray to red to brown
Quantity 800mL
Chemical Name or Material Propylmagnesium chloride
CAS 2234-82-4
CAS 60-29-7
Physical Form Gray to red to brown liquid
Molecular Formula C3H7ClMg
Linear Formula CH3CH2CH2MgCl
Formula Weight 102.85g/mol
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Extremely flammable liquid and vapour.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.
Harmful if swallowed.
May cause drowsiness or dizziness.
Reacts violently with water.
May form explosive peroxides.
Repeated ex
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Extremely flammable liquid and vapour.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.
Harmful if swallowed.
May cause drowsiness or dizziness.
Reacts violently with water.
May form explosive peroxides.
Repeated ex