missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Propylenediphosphonic acid, 98+%

Manufacturer:  Alfa Aesar™ 3027702

 View more versions of this product

Catalog No. AA3027702



Specifications

Specifications

Propylenediphosphonic acid
≥98%
MFCD00050066
DZQCAOQMXPROIJ-UHFFFAOYSA-N
1-phosphonopropan-2-ylphosphonic acid
22023450
Powder
0.5g
4671-82-3
C3H10O6P2
Odorless
CC(CP(=O)(O)O)P(=O)(O)O
204.055
204.06
178°C

Chemical Identifiers

4671-82-3
204.055
DZQCAOQMXPROIJ-UHFFFAOYSA-N
1-phosphonopropan-2-ylphosphonic acid
C3H10O6P2
MFCD00050066
22023450
CC(CP(=O)(O)O)P(=O)(O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

SDS Documents