missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Propylenediphosphonic acid, 98+%

Manufacturer:  Alfa Aesar™ 3027709

 View more versions of this product

Catalog No. AA3027709



Specifications

Specifications

Propylenediphosphonic acid
178°C
4671-82-3
MFCD00050066
DZQCAOQMXPROIJ-UHFFFAOYSA-N
1-phosphonopropan-2-ylphosphonic acid
22023450
Powder
Odorless
10g
C3H10O6P2
propylenediphosphonic acid, acmc-20ajta, propylene-bisphosphonic acid, propylenediphosphonicacid, propane-1,2-diylbis phosphonic acid, 1-phosphonopropan-2-yl phosphonic acid
CC(CP(=O)(O)O)P(=O)(O)O
204.055
204.06
≥98%

Chemical Identifiers

4671-82-3
204.055
DZQCAOQMXPROIJ-UHFFFAOYSA-N
22023450
CC(CP(=O)(O)O)P(=O)(O)O
C3H10O6P2
MFCD00050066
propylenediphosphonic acid, acmc-20ajta, propylene-bisphosphonic acid, propylenediphosphonicacid, propane-1,2-diylbis phosphonic acid, 1-phosphonopropan-2-yl phosphonic acid
1-phosphonopropan-2-ylphosphonic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

SDS Documents