Please login to your online account to display your discounted pricing

Propylene sulfide, 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  346370050

 View more versions of this product

Catalog No. AC346370050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid
Formula Weight 74.14
Quantity 5g
Color Clear colorless
Beilstein 17, II, 15
Density 0.94g/mL
UN Number 1992
Packaging Glass bottle
Molecular Formula C3H6S
Synonym 1, 2-Epithiopropane
Chemical Name or Material Propylene sulfide
Assay Percent Range 96%
CAS 1072-43-1
Health Hazard 1 Danger
Infrared Spectrum Authentic
Refractive Index 1.4740 to 1.4780 (20°C, 589nm)
MDL Number MFCD00005160
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material Propylene sulfide
CAS 1072-43-1
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C3H6S
Formula Weight 74.14
MDL Number MFCD00005160
Synonym 1, 2-Epithiopropane
Accessories

Danger

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

GHS H Statement:
Harmful if inhaled.
Highly flammable liquid and vapor.

SDS & Safety Documents