missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-Propylene oxide, 99+%, Alfa Aesar™

Manufacturer:  Alfa Aesar 30765AE

 View more versions of this product

Catalog No. AA30765AE



Specifications

Specifications

Propylene oxide
100mL
>99%
MFCD00005126
79763
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
CC1CO1
58.08
CHEBI:38685
Liquid
0.83g/mL
75-56-9
C3H6O
UN1280
14,7856
GOOHAUXETOMSMM-UHFFFAOYSA-N
2-methyloxirane
6378
58.08
≥99%

Chemical Identifiers

75-56-9
58.08
GOOHAUXETOMSMM-UHFFFAOYSA-N
6378
2-methyloxirane
C3H6O
MFCD00005126
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
CHEBI:38685
CC1CO1
Safety and Handling

Safety and Handling

GHS H Statement
H224-H340-H350-H302-H312-H332-H315-H319-H335

GHS P Statement
P210-P201-P261-P280-P281-P303+P361+P353-P305+P351+P338-P304+P340-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: PROPYLENE OXIDE

RTECSNumber : TZ2975000

TSCA : TSCA

SDS Documents