Learn More
(±)-Propylene oxide, 99.5%, extra pure, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 430868000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(±)-Propylene oxide | |
75-56-9 | |
100.0 | |
34.0°C | |
Authentic | |
AcroSeal™ Glass Bottle | |
1.3650 to 1.3670 | |
800mL | |
15,7969 | |
Solubility in water: 40g/100mL (20°C). Other solubilities: miscible with alcohol and ether | |
CC1CO1 | |
58.08 | |
0.32 mPa.s (20°C) | |
58.08 | |
Extra Pure |
Extra Pure | |
99.4 | |
0.8300g/mL | |
−37°C | |
99.4% min. (GC) | |
C3H6O | |
MFCD00005126 | |
0.83 | |
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane | |
GOOHAUXETOMSMM-UHFFFAOYNA-N | |
2-methyloxirane | |
6378 | |
CHEBI:38685 | |
99.5% |
Safety and Handling
GHS H Statement
Extremely flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
May cause cancer.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 200-879-2
RUO â Research Use Only