missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-Propylene oxide, 99+%, Alfa Aesar™

Manufacturer:  Alfa Aesar 30765L7

 View more versions of this product

Catalog No. AA30765L7



Specifications

Specifications

(+/-)-Propylene oxide
−37° (−35°F)
1.3660
Colorless
3 x 1L
>99%
MFCD00005126
79763
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
CC1CO1
58.08
CHEBI:38685
Liquid
0.83g/mL
Ether-like
34°C to 35°C
-112°C
75-56-9
C3H6O
UN1280
14,7856
GOOHAUXETOMSMM-UHFFFAOYSA-N
2-methyloxirane
6378
58.08
≥99%

Chemical Identifiers

75-56-9
58.08
GOOHAUXETOMSMM-UHFFFAOYSA-N
6378
2-methyloxirane
C3H6O
MFCD00005126
propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
CHEBI:38685
CC1CO1
Safety and Handling

Safety and Handling

GHS H Statement
H224-H340-H350-H302-H312-H332-H315-H319-H335
Extremely flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: PROPYLENE OXIDE

RTECSNumber : TZ2975000

TSCA : TSCA

SDS Documents