missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propylene Glycol (USP/FCC), Fisher Chemical

Manufacturer:  Thermo Scientific P355200

 View more versions of this product

Catalog No. P355-200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Pass Test (FCC), Pass Test (USP)
200L
57-55-6
4°C max. (FCC)
185° to 189°C (FCC)
≤0.007% (FCC), ≤3.5mg (FCC), 0.007% max. (FCC), 0.007% max. (USP)
Steel Drum
Meets Requirements (USP)
≥99.5% (USP)
≤0.2% (FCC, USP), 0.2% max. (FCC), 0.2% max. (USP)
MFCD00064272
1, 2-Propanediol, 1, 2-Dihydroxypropane, Methyl Glycol
187°C
Undesignated
-60°C
6 to 8
76.095
CHEBI:16997
Viscous Liquid
Propylene Glycol
Vapor Pressure: 0.13mbar at 20°C
≤70ppm (USP), 0.007% max. (USP)
5ppm max. (USP)
Pass Test (FCC), Pass Test (FCC, USP), Pass Test (USP)
1mg/kg max. (FCC)
FCC/USP
1.035 to 1.037 (FCC), 1.035 to 1.037 (FCC, USP), 1.035 to 1.037 (USP)
C3H8O2
CH3CHOHCH2OH
≤60ppm (USP), 0.006% max. (USP)
2.62 (Air = 1.0)
DNIAPMSPPWPWGF-UHFFFAOYSA-N
CC(CO)O
propane-1,2-diol
99°C
1030
76.1

Chemical Identifiers

57-55-6
76.095
DNIAPMSPPWPWGF-UHFFFAOYSA-N
1030
propane-1,2-diol
C3H8O2
MFCD00064272
1, 2-Propanediol, 1, 2-Dihydroxypropane, Methyl Glycol
CHEBI:16997
CC(CO)O
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:0
Flammability:1
Instability:0

SDS Documents