missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propylene Glycol (USP/FCC), Fisher Chemical

Manufacturer:  Fisher Chemical P3551

 View more versions of this product

Catalog No. P355-1




Specifications

Specifications

Pass Test (FCC/USP)
2.62 (Air = 1.0)
Pass Test (FCC, USP)
Glass Bottle
1.035 to 1.037 (FCC, USP)
≤0.2% (FCC, USP)
Undesignated
1L
57-55-6
CH3CHOHCH2OH
1,2-dihydroxypropane, 1,2-propanediol, 1,2-propylene glycol, 2-hydroxypropanol, dowfrost, isopropylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, propylene glycol
CC(CO)O
76.095
CHEBI:16997
Viscous Liquid
FCC/USP
Propylene Glycol
99°C
0.007% max. (FCC/USP)
Meets Requirements (USP)
0.13mbar at 20°C
187°C
-60°C
6 to 8
C3H8O2
MFCD00064272
DNIAPMSPPWPWGF-UHFFFAOYSA-N
propane-1,2-diol
1030
76.1
≥99.5% (USP)

Chemical Identifiers

57-55-6
76.095
DNIAPMSPPWPWGF-UHFFFAOYSA-N
1030
propane-1,2-diol
C3H8O2
MFCD00064272
1,2-dihydroxypropane, 1,2-propanediol, 1,2-propylene glycol, 2-hydroxypropanol, dowfrost, isopropylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, propylene glycol
CHEBI:16997
CC(CO)O
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:0
Flammability:1
Instability:0

SDS Documents