Please login to your online account to display your discounted pricing

Propylene Carbonate, 99.50%, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: fisher scientific acros  131560025

 View more versions of this product

Catalog No. AC131560025

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.4%
Physical Form Liquid
Formula Weight 102.09
Quantity 2.5L
Color Clear
Density 1.2040g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4200 to 1.4220 (20°C, 589nm)
Water 0.1% max.
CAS 108-32-7
Synonym 1, 2-Propanediol cyclic carbonate
Chemical Name or Material Propylene carbonate
Molecular Formula C4H6O3
Assay Percent Range 99.5%
MDL Number MFCD00005385
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Propylene carbonate
CAS 108-32-7
Assay ≥99.4%
Assay Percent Range 99.5%
Molecular Formula C4H6O3
Formula Weight 102.09
MDL Number MFCD00005385
Synonym 1, 2-Propanediol cyclic carbonate
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents