missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propylbenzene, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC418435000

 View more versions of this product

Catalog No. AC418435000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Propylbenzene
103-65-1
Glass bottle
Liquid
98%
C6H5CH2CH2CH3
MFCD00009377
15, 7957
Clear slightly yellow
500g
Authentic
1.4900 to 1.4930 (20°C, 589nm)
≥97.5%
C9H12
120.19
05, 390
0.86g/mL

Chemical Identifiers

98%
C6H5CH2CH2CH3
MFCD00009377
≥97.5%
C9H12
120.19
Propylbenzene

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May be fatal if swallowed and enters airways.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger