missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propylamine Hydroiodide, TCI America™

Manufacturer:  TCI America P22121G

 View more versions of this product

Catalog No. P22121G



Specifications

Specifications

Propylamine Hydroiodide
≥97.0% (N,T)
White
Propylammonium Iodide
CCCN.I
187.024
187.02
14488-45-0
C3H10IN
1g
GIAPQOZCVIEHNY-UHFFFAOYSA-N
propan-1-amine;hydroiodide
91972164
Crystal-Powder at 20°C

Chemical Identifiers

14488-45-0
187.024
Propylammonium Iodide
propan-1-amine;hydroiodide
C3H10IN
GIAPQOZCVIEHNY-UHFFFAOYSA-N
91972164
CCCN.I
Safety and Handling

Safety and Handling

TSCA : No

Documents