missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propylamine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 149312500

 View more versions of this product

Catalog No. AC149312500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
59.11
Clear colorless to pale yellow
0.7190g/mL
1-Aminopropane
Propylamine
98%
Plastic drum
C3H9N
MFCD00008205
Liquid
250mL
Pure
0.5% max. (K.F.)
107-10-8
CH3CH2CH2NH2
1.3870 to 1.3890 (20°C, 589nm)
Authentic
98%

Chemical Identifiers

107-10-8
C3H9N
59.11
1-Aminopropane
≥97.5%
98%
CH3CH2CH2NH2
MFCD00008205
Propylamine, 98%
Pure

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Recommended Storage : Refrigerator +4°C