Please login to your online account to display your discounted pricing

Propyl P-toluenesulfonate 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC380601000

 View more versions of this product

Catalog No. AC380601000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 214.28
Quantity 100g
Color Clear light yellow
Density 1.15g/mL
Packaging Glass bottle
Synonym 4-Methylbenzenesulfonic acid, propyl ester
Chemical Name or Material Propyl p-toluenesulfonate
Infrared Spectrum Authentic
Refractive Index 1.5065 to 1.5085 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C10H14O3S
Linear Formula CH3C6H4SO3(CH2)2CH3
MDL Number MFCD00059244
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 599-91-7
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Propyl p-toluenesulfonate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H14O3S
Linear Formula CH3C6H4SO3(CH2)2CH3
Formula Weight 214.28
MDL Number MFCD00059244
Synonym 4-Methylbenzenesulfonic acid, propyl ester
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents