Please login to your online account to display your discounted pricing

Propyl P-toluenesulfonate 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC380600250

 View more versions of this product

Catalog No. AC380600250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 214.28
Physical Form Liquid
Quantity 25g
Color Clear light yellow
Density 1.15g/mL
Synonym 4-Methylbenzenesulfonic acid, propyl ester
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 599-91-7
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Propyl p-toluenesulfonate
MDL Number MFCD00059244
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5065 to 1.5085 (20°C, 589nm)
Molecular Formula C10H14O3S
Linear Formula CH3C6H4SO3(CH2)2CH3
Assay Percent Range 99%

Specifications

Chemical Name or Material Propyl p-toluenesulfonate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H14O3S
Linear Formula CH3C6H4SO3(CH2)2CH3
Formula Weight 214.28
MDL Number MFCD00059244
Synonym 4-Methylbenzenesulfonic acid, propyl ester
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents