missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propyl Methanesulfonate, TCI America™

Manufacturer:  TCI America P184425G

 View more versions of this product

Catalog No. P184425G



Specifications

Specifications

Propyl Methanesulfonate
Yellow
1912-31-8
MFCD00007561
DKORSYDQYFVQNS-UHFFFAOYSA-N
propyl methanesulfonate
74705
Clear Liquid at 20°C
110°C
25g
C4H10O3S
Methanesulfonic Acid Propyl Ester
CCCOS(=O)(=O)C
138.181
138.18
≥98.0% (GC)

Chemical Identifiers

1912-31-8
138.181
DKORSYDQYFVQNS-UHFFFAOYSA-N
74705
CCCOS(=O)(=O)C
C4H10O3S
MFCD00007561
Methanesulfonic Acid Propyl Ester
propyl methanesulfonate
Safety and Handling

Safety and Handling

RTECSNumber : PB2680000

TSCA : No

Documents