missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propyl disulfide, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 232340050

 View more versions of this product

Catalog No. AC232340050



Specifications

Specifications

Propyl disulfide
66°C
Glass bottle
0.95
Colorless to Yellow
5g
629-19-6
C6H14S2
MFCD00009378
Solubility in water: insoluble.
CCCSSCCC
150.24
CHEBI:45758
Liquid
0.9500g/mL
Authentic
1.4940 to 1.5000
195.0°C to 196.0°C
-86.0°C
99%
98.5% min. (GC)
[CH3(CH2)2S]2
1,1'-dithiodipropane, 1-propyldisulfanyl propane, 4,5-dithiaoctane, di-n-propyl disulfide, dipropyl disulfide, dipropyl disulphide, disulfide, dipropyl, n-propyl disulfide, propyl disulfide, propyldithiopropane
ALVPFGSHPUPROW-UHFFFAOYSA-N
1-(propyldisulfanyl)propane
12377
150.24
99%

Chemical Identifiers

629-19-6
150.24
ALVPFGSHPUPROW-UHFFFAOYSA-N
12377
1-(propyldisulfanyl)propane
C6H14S2
MFCD00009378
1,1'-dithiodipropane, 1-propyldisulfanyl propane, 4,5-dithiaoctane, di-n-propyl disulfide, dipropyl disulfide, dipropyl disulphide, disulfide, dipropyl, n-propyl disulfide, propyl disulfide, propyldithiopropane
CHEBI:45758
CCCSSCCC
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-079-8

RTECSNumber : JO1955000

TSCA : TSCA

SDS Documents