Please login to your online account to display your discounted pricing

Propyl disulfide, 99%, ACROS Organics™

$32.78 - $32.78

Chemical Identifiers

Chemical Name or Material Propyl disulfide
Name Note 99%
CAS 629-19-6
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H14S2
Linear Formula {CH3(CH2)2S}2
Formula Weight 150.24
MDL Number MFCD00009378
Synonym Di-n-propyl disulfide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC232340050

  SDS
thermo fisher scientific
232340050
Glass bottle 5g Each for $32.78
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 150.24
Physical Form Liquid
Quantity 5g
Color Clear colorless to pale yellow
TSCA TSCA
RTECS Number JO1955000
EINECS Number 211-079-8
Density 0.95g/mL
Health Hazard 1 7
Synonym Di-n-propyl disulfide
Infrared Spectrum Authentic
Refractive Index 1.4940 to 1.5000 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Name Note 99%
Chemical Name or Material Propyl disulfide
Molecular Formula C6H14S2
Linear Formula {CH3(CH2)2S}2
CAS 629-19-6
Assay Percent Range 99%
MDL Number MFCD00009378
Safety and Handling

7

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.