Please login to your online account to display your discounted pricing

Propyl 4-hydroxybenzoate, 99+%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  131595000

 View more versions of this product

Catalog No. AC131595000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 180.2
Quantity 500g
Color White
Beilstein 10, 160
Merck Index 15, 7977
EINECS Number 202-307-7
TSCA TSCA
RTECS Number DH2800000
Melting Point 95° to 99°C
Chemical Name or Material Propyl 4-hydroxybenzoate
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (5 hrs) (over silica gel)
Packaging Plastic bottle
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C10H12O3
Linear Formula HOC6H4CO2CH2CH2CH3
Sulfated Ash 0.1% max.
MDL Number MFCD00002354
CAS 94-13-3
Synonym Propylparaben
Appearance of Solution (5% in ethanol) clear colorless
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Propyl 4-hydroxybenzoate
Name Note 99+%
CAS 94-13-3
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C10H12O3
Linear Formula HOC6H4CO2CH2CH2CH3
Formula Weight 180.2
MDL Number MFCD00002354
Synonym Propylparaben
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents