Learn More
Propyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131591000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Propyl 4-hydroxybenzoate | |
94-13-3 | |
White | |
0.5% max. (5 hrs) (over silica gel) | |
99% min. (GC) | |
C10H12O3 | |
MFCD00002354 | |
10, 160 | |
propylparaben, propyl paraben, 4-hydroxybenzoic acid propyl ester, propyl p-hydroxybenzoate, nipasol, nipazol, propyl butex, betacide p, propyl parahydroxybenzoate, propylparasept | |
QELSKZZBTMNZEB-UHFFFAOYSA-N | |
propyl 4-hydroxybenzoate | |
7175 | |
180.2 | |
Crystalline Powder |
99+% | |
95°C to 99°C | |
180°C | |
Authentic | |
Plastic bottle | |
HOC6H4CO2CH2CH2CH3 | |
100 g | |
15, 7977 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether and acetone | |
CCCOC(=O)C1=CC=C(C=C1)O | |
180.2 | |
CHEBI:32063 | |
99+% |
Chemical Identifiers
94-13-3 | |
180.2 | |
QELSKZZBTMNZEB-UHFFFAOYSA-N | |
7175 | |
propyl 4-hydroxybenzoate |
C10H12O3 | |
MFCD00002354 | |
propylparaben, propyl paraben, 4-hydroxybenzoic acid propyl ester, propyl p-hydroxybenzoate, nipasol, nipazol, propyl butex, betacide p, propyl parahydroxybenzoate, propylparasept | |
CHEBI:32063 | |
CCCOC(=O)C1=CC=C(C=C1)O |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 202-307-7
RTECSNumber : DH2800000
TSCA : TSCA
RUO – Research Use Only