Please login to your online account to display your discounted pricing
Propper Velket™ Tourniquet
Flexible rubber band is easily applied to any limb
$14.90 - $14.90
Products
Catalog Number Mfr. No. Price Quantity    
15-234
propper manufacturing co
221010
Each for $14.90
Pack of 12 Each for $178.80
Description & Specifications
  • Induces venous or arterial constriction for maximum ease in drawing blood
  • Eliminates rubber tube “pinch” and danger of ecchymosis due to extravasation
  • Features a Velcro™ fastener that holds firmly and securely at any desired degree of tension
  • A single tug removes the tourniquet from the patient
  • Packaged in a plastic box with illustrations