missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Propiophenone, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 131540025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Propiophenone | |
1.0000g/mL | |
87°C | |
98.5% min. (GC) | |
C9H10O | |
C6H5COC2H5 | |
07,II,231 | |
15,7944 | |
Solubility in water: insoluble. Other solubilities: miscible with methanol, ether, benzene and toluene, insoluble in glycerol, ethylene glycol and, propylene glycol | |
CCC(=O)C1=CC=CC=C1 | |
134.18 | |
CHEBI:425902 | |
99% |
93-55-0 | |
218.0°C | |
Authentic | |
Glass bottle | |
1.5248 to 1.5268 | |
2.5L | |
1 | |
propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one | |
KRIOVPPHQSLHCZ-UHFFFAOYSA-N | |
1-phenylpropan-1-one | |
7148 | |
134.18 |
Safety and Handling
EINECSNumber : 202-257-6
RUO â Research Use Only