missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Propionophenone p-toluenesulfonylhydrazone, 97%

Manufacturer:  Alfa Aesar™ H5340914

 View more versions of this product

Catalog No. AAH5340914



Specifications

Specifications

Propionophenone p-toluenesulfonylhydrazone
25g
C16H18N2O2S
1-propiophenone tosylhydrazone, 4-methyl-n'-1-phenylpropylidene benzenesulfonohydrazide, 1-propiophenone tosylhydrazone, 1-propiophenone-p-toluenesulfonylhydrazone, propionophenone p-toluenesulfonylhydrazone, 4-methyl-n-e-1-phenylpropylideneamino benzenesulfonamide, 1e-2-phenyl-1-azabut-1-enyl 4-methylphenyl sulfonyl amine, 4-methyl-n'-1z-1-phenylpropylidene benzenesulfonohydrazide
CCC(=NNS(=O)(=O)C1=CC=C(C=C1)C)C2=CC=CC=C2
302.392
302.39
118°C to 120°C
17336-66-2
MFCD00159336
TWTWWQDWKPRWFI-WUKNDPDISA-N
4-methyl-N-[(E)-1-phenylpropylideneamino]benzenesulfonamide
6374388
97%

Chemical Identifiers

17336-66-2
302.392
TWTWWQDWKPRWFI-WUKNDPDISA-N
6374388
CCC(=NNS(=O)(=O)C1=CC=C(C=C1)C)C2=CC=CC=C2
C16H18N2O2S
MFCD00159336
1-propiophenone tosylhydrazone, 4-methyl-n'-1-phenylpropylidene benzenesulfonohydrazide, 1-propiophenone tosylhydrazone, 1-propiophenone-p-toluenesulfonylhydrazone, propionophenone p-toluenesulfonylhydrazone, 4-methyl-n-e-1-phenylpropylideneamino benzenesulfonamide, 1e-2-phenyl-1-azabut-1-enyl 4-methylphenyl sulfonyl amine, 4-methyl-n'-1z-1-phenylpropylidene benzenesulfonohydrazide
4-methyl-N-[(E)-1-phenylpropylideneamino]benzenesulfonamide
SDS Documents