Learn More
Propionic acid, sodium salt, 99.0-100.5%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 149010010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Propionic acid, sodium salt | |
285°C to 286°C | |
>250°C | |
99.0-100.5% | |
C3H5NaO2 | |
MFCD00002759 | |
02, 234 | |
sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar | |
JXKPEJDQGNYQSM-UHFFFAOYSA-M | |
sodium;propanoate | |
2723816 | |
99.0-100.5% |
137-40-6 | |
White | |
Authentic | |
Plastic bottle | |
C2H5CO2Na | |
1 kg | |
15, 8798 | |
Solubility in water: very soluble. Other solubilities: soluble in alcohol | |
CCC(=O)[O-].[Na+] | |
96.06 | |
96.06 | |
Crystalline Powder |
Chemical Identifiers
137-40-6 | |
96.06 | |
JXKPEJDQGNYQSM-UHFFFAOYSA-M | |
2723816 | |
CCC(=O)[O-].[Na+] |
C3H5NaO2 | |
MFCD00002759 | |
sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar | |
sodium;propanoate |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge
EINECSNumber : 205-290-4
RUO – Research Use Only