Please login to your online account to display your discounted pricing

Propionic acid, 99+%, extra pure, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC220130025

 View more versions of this product

Catalog No. AC220130025

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 74.08
Purity Grade Extra Pure
Quantity 2.5L
Color Clear
Density 0.99g/mL
Beilstein 02, 234
Merck Index 15, 7939
Fieser 12,415
Refractive Index 1.3850 to 1.3870 (20°C, 589nm)
Linear Formula C2H5CO2H
Infrared Spectrum Authentic
Chemical Name or Material Propionic acid
Packaging Glass bottle
Molecular Formula C3H6O2
CAS 79-09-4
Assay Percent Range 99+%
Health Hazard 1 GHS Signal Word: Danger
Water 0.1% max.
MDL Number MFCD00002756
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Propionic acid
CAS 79-09-4
Purity Grade Extra Pure
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C3H6O2
Linear Formula C2H5CO2H
Formula Weight 74.08
MDL Number MFCD00002756
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapour.

SDS & Safety Documents