missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Propionic acid, ≥99.5%, Honeywell™ Fluka™

≥99.5%

Manufacturer:  Honeywell Fluka™ P13861L

 View more versions of this product

Catalog No. 60048112



Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Propionic acid
54°C (129.2°F)
Unpleasant
2.55 (vs air)
141°C (1013hPa)
-21°C
79-09-4
CH3CH2COOH
UN3463
carboxyethane
XBDQKXXYIPTUBI-UHFFFAOYSA-N
propanoic acid
1032
74.08g/mol
≥99.5%
0.993g/mL at 25°C
955°F
Glass bottle
2.4mmHg (20°C)
Undesignated
1L
C3H6O2
MFCD00002756
506071
Completely miscible in water
CCC(=O)O
74.079
CHEBI:30768
Liquid

Chemical Identifiers

79-09-4
74.079
XBDQKXXYIPTUBI-UHFFFAOYSA-N
1032
propanoic acid
C3H6O2
MFCD00002756
carboxyethane
CHEBI:30768
CCC(=O)O
Safety and Handling

Safety and Handling

P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P235

DOTInformation : class 3 - Propionic acid

RTECSNumber : 201-176-3

Documents