missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Propionamidoxime, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5088903
Description
Propionamidoxime is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPropionamidoxime is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
It is hygroscopic. Incompatible with oxidizing agents.
Specifications
Propionamidoxime | |
C3H8N2O | |
1 g | |
n-hydroxypropionamidine, n-hydroxypropionimidamide, 1z-n'-hydroxypropanimidamide, 1-hydroxyimino propylamine, n-hydroxy-propionamidine, n-hydroxyproponmdamde, n-hydroxypropanamidine, n'-hydroxypropimidamide, z-n'-hydroxypropimidamide, e-n'-hydroxypropanimidamide | |
RLZPCFQNZGINRP-UHFFFAOYSA-N | |
N'-hydroxypropanimidamide | |
6521662 | |
97% |
29335-36-2 | |
MFCD03426272 | |
Hygroscopic | |
Slightly soluble in water. | |
CCC(=NO)N | |
88.11 | |
88.11 |
Chemical Identifiers
29335-36-2 | |
88.11 | |
RLZPCFQNZGINRP-UHFFFAOYSA-N | |
6521662 | |
CCC(=NO)N |
C3H8N2O | |
MFCD03426272 | |
n-hydroxypropionamidine, n-hydroxypropionimidamide, 1z-n'-hydroxypropanimidamide, 1-hydroxyimino propylamine, n-hydroxy-propionamidine, n-hydroxyproponmdamde, n-hydroxypropanamidine, n'-hydroxypropimidamide, z-n'-hydroxypropimidamide, e-n'-hydroxypropanimidamide | |
N'-hydroxypropanimidamide |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only