Please login to your online account to display your discounted pricing

Alfa Aesar™ Propionamide, 98%

$172.41 - $698.84

Chemical Identifiers

Chemical Name or Material Propionamide
CAS 79-05-0
Molecular Formula CH3CH2CONH2
Formula Weight 73.1
MDL Number MFCD00008039
Merck Index 14,7824
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1052122

  SDS
alfa aesar™
A1052122
100g Each for $172.41

AAA1052136

  SDS
alfa aesar™
A1052136
500g Each for $698.84
Description & Specifications

Specifications

Formula Weight 73.1
Chemical Name or Material Propionamide
TSCA TSCA
Assay 98%
Molecular Formula CH3CH2CONH2
MDL Number MFCD00008039
CAS 79-05-0
Merck Index 14,7824
Boiling Point 213°C
Melting Point 76° to 79°C
Quantity 100g
RTECS Number UE2975000

Applications
Propionamide was used as adsorbent in the determination of adsorption isotherms of acetamide and propionamide on multi-wall carbon nanotube1. It was used in a robust screening method to study biotransformations using (+)-γ-lactamase enzyme. Propionamide on γ-irradiation reacts with sulfur dioxide and this reaction has been studied by ESR spectroscopy, gas absorption measurements and X-ray diffraction3

Solubility
Soluble in water, chloroform, ethanol, diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids, bases, strong reducing agents.