Please login to your online account to display your discounted pricing

Propionaldehyde, 99+%, ACROS Organics™


Quantity 10L
Packaging Plastic drum

Manufacturer: thermo fisher scientific  AC220510100

 View more versions of this product

Catalog No. AC220510100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Platelets
Assay ≥99.0% (GC)
Formula Weight 58.08
Quantity 10L
Color White
EINECS Number 204-623-0
TSCA TSCA
Density 0.7980g/mL
RTECS Number UE0350000
Infrared Spectrum Authentic
Packaging Plastic drum
Refractive Index 1.3610 to 1.3660 (20°C, 589nm)
Water 0.5% max. (K.F.)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym Propanal
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Chemical Name or Material Propionaldehyde
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C3H6O
Linear Formula C2H5CHO
MDL Number MFCD00007020
CAS 123-38-6

Specifications

Chemical Name or Material Propionaldehyde
Name Note 99+%
CAS 123-38-6
Assay ≥99.0% (GC)
Assay Percent Range 99+%
Molecular Formula C3H6O
Linear Formula C2H5CHO
Formula Weight 58.08
MDL Number MFCD00007020
Synonym Propanal
Accessories

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.