Learn More
Propionaldehyde, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131510250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Propionaldehyde | |
-81.0°C | |
0.7980g/mL | |
−40°C | |
96% min. (GC) | |
C3H6O | |
C2H5CHO | |
01,629 | |
15,7937 | |
Solubility in water: 540g/L (20°C). Other solubilities: soluble in alcohol and ether | |
CCC=O | |
58.08 | |
0.6 mPa.s (20°C) | |
58.08 | |
Liquid |
123-38-6 | |
Colorless | |
48.0°C | |
Authentic | |
Glass bottle | |
1.3610 to 1.3650 | |
25 mL | |
0.798 | |
propionaldehyde, propanaldehyde, propaldehyde, propional, propionic aldehyde, n-propanal, methylacetaldehyde, propylaldehyde, propylic aldehyde, aldehyde propionique | |
NBBJYMSMWIIQGU-UHFFFAOYSA-N | |
propanal | |
527 | |
CHEBI:17153 | |
97% |
Chemical Identifiers
123-38-6 | |
58.08 | |
propionaldehyde, propanaldehyde, propaldehyde, propional, propionic aldehyde, n-propanal, methylacetaldehyde, propylaldehyde, propylic aldehyde, aldehyde propionique | |
CHEBI:17153 | |
CCC=O |
C3H6O | |
NBBJYMSMWIIQGU-UHFFFAOYSA-N | |
527 | |
propanal |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 204-623-
RUO – Research Use Only