missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Propiolaldehyde diethyl acetal, 97%

Manufacturer:  Alfa Aesar™ L1076714

 View more versions of this product

Catalog No. AAL1076714



Description

Description

Applications
Propiolaldehyde diethyl acetal is used in preparation of 3-boronoacrolein pinacolate, a heterodyne and (E)-3-(tributylstannyl)-2-propenal.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Air and moisture sensitive. Store under inert gas.
Specifications

Specifications

Propiolaldehyde diethyl acetal
138°C to 139°C
10160-87-9
MFCD00009237
1701566
RGUXEWWHSQGVRZ-UHFFFAOYSA-N
3,3-diethoxyprop-1-yne
66285
97%
32°C (89°F)
25g
C7H12O2
UN1993
3,3-diethoxy-1-propyne, propargylaldehyde diethyl acetal, 3,3-diethoxypropyne, propiolaldehyde diethyl acetal, 1-propyne, 3,3-diethoxy, 3,3-diethoxy-propyne, propynal diethyl acetal, 1,1-diethoxyprop-2-yne, 1-propyne,3,3-diethoxy, propargyl aldehyde diethylacetal
CCOC(C#C)OCC
128.171
128.17

Chemical Identifiers

10160-87-9
128.171
RGUXEWWHSQGVRZ-UHFFFAOYSA-N
66285
CCOC(C#C)OCC
C7H12O2
MFCD00009237
3,3-diethoxy-1-propyne, propargylaldehyde diethyl acetal, 3,3-diethoxypropyne, propiolaldehyde diethyl acetal, 1-propyne, 3,3-diethoxy, 3,3-diethoxy-propyne, propynal diethyl acetal, 1,1-diethoxyprop-2-yne, 1-propyne,3,3-diethoxy, propargyl aldehyde diethylacetal
3,3-diethoxyprop-1-yne
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Recommended Storage : 2° to 8°C

Documents