Please login to your online account to display your discounted pricing

Alfa Aesar™ Propidium iodide,/ml aqueous soln.


Quantity 10mL

Manufacturer: alfa aesar™  J66584AB

 View more versions of this product

Catalog No. AAJ66584AB

  • / Each
 

Description & Specifications

Chemical Identifiers

MDL Number MFCD00011921
Chemical Name or Material Propidium iodide
Physical Form Liquid
Beilstein 3843838
Synonym 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide; Phenanthridinium
Color Orange-red
Quantity 10mL
Sensitivity Light Sensitive
Name Note 1mg/mL aqueous soln.
Storage Note 1 2° to 8°C
RTECS Number SF7949600

Specifications

Chemical Name or Material Propidium iodide
Name Note 1mg/mL aqueous soln.
MDL Number MFCD00011921
Beilstein 3843838
Synonym 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide; Phenanthridinium

Applications
Membrane impermeant, phenanthridinium intercalator dye excited by mercury or xenon-arc lamps or argon-ion laser. Selectively stains dead cells and a counter stain for multicolor fluorescent images.

Solubility
Difficult to mix.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Accessories

2° to 8°C

SDS & Safety Documents