Please login to your online account to display your discounted pricing

Alfa Aesar™ Propidium iodide,/ml aqueous soln.

$105.32 - $105.32

Chemical Identifiers

Chemical Name or Material Propidium iodide
Name Note 1mg/mL aqueous soln.
MDL Number MFCD00011921
Beilstein 3843838
Synonym 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide; Phenanthridinium
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66584AB

  SDS
alfa aesar™
J66584AB
10mL Each for $105.32
Description & Specifications

Specifications

MDL Number MFCD00011921
Chemical Name or Material Propidium iodide
Color Orange-red
Synonym 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide; Phenanthridinium
Quantity 10mL
Physical Form Liquid
Beilstein 3843838
Sensitivity Light Sensitive
Storage Note 1 2° to 8°C
Name Note 1mg/mL aqueous soln.
RTECS Number SF7949600

Applications
Membrane impermeant, phenanthridinium intercalator dye excited by mercury or xenon-arc lamps or argon-ion laser. Selectively stains dead cells and a counter stain for multicolor fluorescent images.

Solubility
Difficult to mix.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

2° to 8°C