Please login to your online account to display your discounted pricing

Propargyltrimethylsilane, 80-90%, stabilized, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC366630010

 View more versions of this product

Catalog No. AC366630010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 112.25
Quantity 1g
Color Clear yellow
Beilstein 04, IV, 3938
Density 0.7500g/mL
Assay 80 to 90% (typically) exact purity on the certificate of analysis
Infrared Spectrum Authentic
Molecular Formula C6H12Si
Linear Formula HCidentical toCCH2Si(CH3)3
Packaging Glass bottle
MDL Number MFCD00042922
CAS 13361-64-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Freezer -20°C
Chemical Name or Material Propargyltrimethylsilane
Name Note 90%
Assay Percent Range 80-90%
Stabilizer 0.1 wt% hydroquinone

Specifications

Chemical Name or Material Propargyltrimethylsilane
Name Note 90%
CAS 13361-64-3
Assay 80 to 90% (typically) exact purity on the certificate of analysis
Assay Percent Range 80-90%
Molecular Formula C6H12Si
Linear Formula HCidentical toCCH2Si(CH3)3
Formula Weight 112.25
MDL Number MFCD00042922
Accessories

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

Freezer -20°C

SDS & Safety Documents