Please login to your online account to display your discounted pricing

Propargyloxytrimethylsilane, 97%, ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC313900025

 View more versions of this product

Catalog No. AC313900025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 128.25
Physical Form Liquid
Quantity 2.5g
Color Clear yellow-brown
EINECS Number 226-981-7
TSCA TSCA
Density 0.8300g/mL
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Refractive Index 1.4070 to 1.4090 (20°C, 589nm)
Synonym Trimethyl(2-propynyloxy)silane
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 97%
Chemical Name or Material Propargyloxytrimethylsilane
Assay Percent Range 97%
Molecular Formula C6H12OSi
Linear Formula HCidentical toCCH2OSi(CH3)3
CAS 5582-62-7
MDL Number MFCD00042844

Specifications

Chemical Name or Material Propargyloxytrimethylsilane
Name Note 97%
CAS 5582-62-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H12OSi
Linear Formula HCidentical toCCH2OSi(CH3)3
Formula Weight 128.25
MDL Number MFCD00042844
Synonym Trimethyl(2-propynyloxy)silane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.

SDS & Safety Documents