Please login to your online account to display your discounted pricing

Propargyloxytrimethylsilane, 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  313900100

 View more versions of this product

Catalog No. AC313900100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 128.25
Quantity 10g
Color Clear yellow-brown
EINECS Number 226-981-7
TSCA TSCA
Density 0.8300g/mL
Name Note 97%
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C6H12OSi
Packaging Glass bottle
MDL Number MFCD00042844
CAS 5582-62-7
Refractive Index 1.4070 to 1.4090 (20°C, 589nm)
Synonym Trimethyl(2-propynyloxy)silane
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Propargyloxytrimethylsilane
Linear Formula HCidentical toCCH2OSi(CH3)3

Specifications

Chemical Name or Material Propargyloxytrimethylsilane
Name Note 97%
CAS 5582-62-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H12OSi
Linear Formula HCidentical toCCH2OSi(CH3)3
Formula Weight 128.25
MDL Number MFCD00042844
Synonym Trimethyl(2-propynyloxy)silane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.

SDS & Safety Documents