Learn More
Propargylamine hydrochloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131470100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Propargylamine hydrochloride | |
175°C to 182°C | |
Glass bottle | |
HC∽CCH2NH2·HCl | |
10 g | |
propargylamine hydrochloride, prop-2-yn-1-amine hydrochloride, 2-propynylamine hydrochloride, propargylamine hcl, 2-propynylamine hcl, 3-amino-1-propyne hydrochloride, prop-2-ynylamine hydrochloride, 2-propyn-1-amine, hydrochloride, pubchem18845, acmc-1bvqh | |
[H+].[Cl-].NCC#C | |
91.54 | |
91.54 |
15430-52-1 | |
94% min. (Argentometry) | |
C3H6ClN | |
MFCD00012907 | |
04, 228 | |
IKXNIQJDNKPPCH-UHFFFAOYSA-N | |
prop-2-yn-1-amine;hydrochloride | |
11205720 | |
95% |
Chemical Identifiers
15430-52-1 | |
91.54 | |
IKXNIQJDNKPPCH-UHFFFAOYSA-N | |
11205720 | |
[H+].[Cl-].NCC#C |
C3H6ClN | |
MFCD00012907 | |
propargylamine hydrochloride, prop-2-yn-1-amine hydrochloride, 2-propynylamine hydrochloride, propargylamine hcl, 2-propynylamine hcl, 3-amino-1-propyne hydrochloride, prop-2-ynylamine hydrochloride, 2-propyn-1-amine, hydrochloride, pubchem18845, acmc-1bvqh | |
prop-2-yn-1-amine;hydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
EINECSNumber : 239-440-5
RUO – Research Use Only