missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Propargyl Sulfide 98.0+%, TCI America™
$353.37 - $1058.44
Chemical Identifiers
CAS | 13702-09-5 |
---|---|
Molecular Formula | C6H6S |
Molecular Weight (g/mol) | 110.174 |
MDL Number | MFCD00048109 |
InChI Key | XPTMJJIPRSWBDK-UHFFFAOYSA-N |
Synonym | Dipropargyl Sulfide |
PubChem CID | 518812 |
IUPAC Name | 3-prop-2-ynylsulfanylprop-1-yne |
SMILES | C#CCSCC#C |
Chemical Identifiers
13702-09-5 | |
110.174 | |
XPTMJJIPRSWBDK-UHFFFAOYSA-N | |
518812 | |
C#CCSCC#C |
C6H6S | |
MFCD00048109 | |
Dipropargyl Sulfide | |
3-prop-2-ynylsulfanylprop-1-yne |
Specifications
13702-09-5 | |
C6H6S | |
1 g | |
Dipropargyl Sulfide | |
C#CCSCC#C | |
110.174 | |
110.17 | |
Liquid |
59°C | |
MFCD00048109 | |
1993 | |
XPTMJJIPRSWBDK-UHFFFAOYSA-N | |
3-prop-2-ynylsulfanylprop-1-yne | |
518812 | |
≥98.0% (GC) | |
Propargyl Sulfide |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator