Please login to your online account to display your discounted pricing

Alfa Aesar™ Propargyl propionate, 98%

Chemical Identifiers

Chemical Name or Material Propargyl propionate
CAS 1932-92-9
Molecular Formula CH3CH2CO2CH2C≡CH
Formula Weight 112.13
MDL Number MFCD00039917
UN Number UN3272
Beilstein 1902510
Synonym Propionic acid propargyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0937109

  SDS
alfa aesar™
L0937109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0937118

  SDS
alfa aesar™
L0937118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.13
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Propargyl propionate
Health Hazard 2 GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
UN Number UN3272
Assay 98%
CAS 1932-92-9
Molecular Formula CH3CH2CO2CH2C≡CH
Health Hazard 1 Danger
MDL Number MFCD00039917
Synonym Propionic acid propargyl ester
Flash Point 40°C (104°F)
Beilstein 1902510
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Boiling Point 137° to 139°C
Quantity 10g
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.