Please login to your online account to display your discounted pricing

Propargyl p-toluenesulfonate, 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC379681000

 View more versions of this product

Catalog No. AC379681000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 210.25
Physical Form Liquid
Quantity 100g
Color Clear colorless to brown
Beilstein 11, IV, 263
Packaging Glass bottle
Linear Formula CH3C6H4SO3CH2CCH
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C10H10O3S
MDL Number MFCD01462194
Assay Percent Range 97%
Refractive Index 1.5270 to 1.5290 (20°C, 589nm)
Chemical Name or Material Propargyl p-toluenesulfonate
CAS 6165-76-0
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Propargyl p-toluenesulfonate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H10O3S
Linear Formula CH3C6H4SO3CH2CCH
Formula Weight 210.25
MDL Number MFCD01462194
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents