Please login to your online account to display your discounted pricing

Alfa Aesar™ Propargyl p-toluenesulfonate, 97%

$100.68 - $100.68

Chemical Identifiers

Chemical Name or Material Propargyl p-toluenesulfonate
CAS 6165-76-0
Molecular Formula C10H10O3S
Formula Weight 210.25
MDL Number MFCD00078365
Beilstein 1212957
Synonym p-Toluenesulfonic acid propargyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1207509

 
alfa aesar™
L1207509
10g Each for $100.68
Description & Specifications

Specifications

Formula Weight 210.25
Chemical Name or Material Propargyl p-toluenesulfonate
Assay 97%
MDL Number MFCD00078365
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C10H10O3S
Beilstein 1212957
Boiling Point 117° to 121°C (0.3mmHg)
Quantity 10g
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
RTECS Number XT7600000
CAS 6165-76-0
Synonym p-Toluenesulfonic acid propargyl ester

Applications
Propargyl p-toluenesulfonate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in Chloroform, Dichloromethane, DMSO, Ethyl Acetate. Not miscible in water.

Notes
Store at 4°C. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.