missing translation for 'onlineSavingsMsg'
Learn More

Propargyl methacrylate, 98%, stab. with 200-300ppm 4-methoxyphenol

CAS: 13861-22-8 | C7H8O2 | 124.14 g/mol

$218.90 - $880.89

Chemical Identifiers

CAS 13861-22-8
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00048123
InChI Key PZAWASVJOPLHCJ-UHFFFAOYSA-N
Synonym propargyl methacrylate, 2-propynyl methacrylate, 2-propenoic acid, 2-methyl-, 2-propynyl ester, 2-propenoic acid, 2-methyl-, 2-propyn-1-yl ester, prop-2-yn-1-yl 2-methylprop-2-enoate, prop-2-ynyl methacrylate, propargylmethacrylate, acmc-1bvgj, prop-2-yn-1-yl methacrylate, propargyl methacrylate, stabilized 10g
PubChem CID 83778
IUPAC Name prop-2-ynyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1155609
View Documents
Thermo Scientific Chemicals
L1155609
10 g
Each for $218.90
Only null left
 
AAL1155618
View Documents
Thermo Scientific Chemicals
L1155618
50 g
Each for $880.89
Only null left
 
Description

Description

Propargyl methacrylate is a monomer used in polymerization reactions. It is associated with its acetylene and under goes polymerization reaction through the reversible addition fragmentation chain transfer (RAFT) process using cyanoisopropyl dithiobenzoate as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl methacrylate is a monomer used in polymerization reactions. It is associated with its acetylene and under goes polymerization reaction through the reversible addition fragmentation chain transfer (RAFT) process using cyanoisopropyl dithiobenzoate as a reagent.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

13861-22-8
124.14
PZAWASVJOPLHCJ-UHFFFAOYSA-N
83778
CC(=C)C(=O)OCC#C
C7H8O2
MFCD00048123
propargyl methacrylate, 2-propynyl methacrylate, 2-propenoic acid, 2-methyl-, 2-propynyl ester, 2-propenoic acid, 2-methyl-, 2-propyn-1-yl ester, prop-2-yn-1-yl 2-methylprop-2-enoate, prop-2-ynyl methacrylate, propargylmethacrylate, acmc-1bvgj, prop-2-yn-1-yl methacrylate, propargyl methacrylate, stabilized 10g
prop-2-ynyl 2-methylprop-2-enoate

Specifications

13861-22-8
149°C to 151°C
C7H8O2
MFCD00048123
UN3272
propargyl methacrylate, 2-propynyl methacrylate, 2-propenoic acid, 2-methyl-, 2-propynyl ester, 2-propenoic acid, 2-methyl-, 2-propyn-1-yl ester, prop-2-yn-1-yl 2-methylprop-2-enoate, prop-2-ynyl methacrylate, propargylmethacrylate, acmc-1bvgj, prop-2-yn-1-yl methacrylate, propargyl methacrylate, stabilized 10g
PZAWASVJOPLHCJ-UHFFFAOYSA-N
prop-2-ynyl 2-methylprop-2-enoate
83778
98%
0.986
48°C (118°F)
1.45
10 g
906850
Immiscible with water.
CC(=C)C(=O)OCC#C
124.14
124.14
Propargyl methacrylate, stabilized with 250ppm 4-methoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 237-599-5

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only