missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propargyl ether, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5347314

 View more versions of this product

Catalog No. AAH5347314


Description

Description

Propargyl ether is a useful ether for proteomics research. It was used for the preparation of polyferrocenes, via polyaddition with 1,1?-bis(azidoethyl)ferrocene. It may be used for the preaparation of cyclic polystyrene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl ether is a useful ether for proteomics research. It was used for the preparation of polyferrocenes, via polyaddition with 1,1′-bis(azidoethyl)ferrocene. It may be used for the preaparation of cyclic polystyrene.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

Propargyl ether
0.914
24°C (75°F)
1.443
MFCD00048108
UN3271
HRDCVMSNCBAMAM-UHFFFAOYSA-N
3-prop-2-ynoxyprop-1-yne
23349
98%
6921-27-3
55°C to 57°C (65 mmHg)
C6H6O
(HC≡CCH2)2O
25 g
propargyl ether, dipropargyl ether, 2-propynyl ether, 1-propyne, 3,3'-oxybis, bispropargyl ether, 3-prop-2-yn-1-yloxy prop-1-yne, propargyl-ether, dipropargyl oxide, di 2-propynyl ether, bis 2-propynyl ether
C#CCOCC#C
94.113
94.11

Chemical Identifiers

6921-27-3
94.113
HRDCVMSNCBAMAM-UHFFFAOYSA-N
23349
C#CCOCC#C
C6H6O
MFCD00048108
propargyl ether, dipropargyl ether, 2-propynyl ether, 1-propyne, 3,3'-oxybis, bispropargyl ether, 3-prop-2-yn-1-yloxy prop-1-yne, propargyl-ether, dipropargyl oxide, di 2-propynyl ether, bis 2-propynyl ether
3-prop-2-ynoxyprop-1-yne
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c

H226-H302

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

RTECSNumber : UK5950000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only