Please login to your online account to display your discounted pricing

Alfa Aesar™ Propargyl chloride, 70% w/w soln. in toluene

Chemical Identifiers

Chemical Name or Material Propargyl chloride
CAS 624-65-7
Molecular Formula C3H3Cl
Linear Formula HC≡CCH2Cl
Formula Weight 74.51
MDL Number MFCD00000980
UN Number UN2924
Beilstein 506005
Merck Index 14,7810
Synonym 3-Chloro-1-propyne
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2041614

 
alfa aesar™
L2041614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2041622

 
alfa aesar™
L2041622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 74.51
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Melting Point <−50°C
Quantity 25g
Concentration 70% w/w soln. in toluene
Linear Formula HC≡CCH2Cl
MDL Number MFCD00000980
CAS 624-65-7
Beilstein 506005
Merck Index 14,7810
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H301-H311-H331-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Boiling Point 60° to 63°C
Molecular Formula C3H3Cl
Synonym 3-Chloro-1-propyne
Flash Point −15°C (−5°F)
UN Number UN2924
Chemical Name or Material Propargyl chloride
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H301-H311-H331-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.