missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Propargyl benzoate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5344406
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Propargyl benzoate | |
1.106 | |
C9H12N3Na2O8P | |
1.532 | |
5 g | |
INTPYBRGLGSMRA-IJRYJGAHNA-L | |
disodium;[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate | |
94332 | |
98% |
6750-04-5 | |
225°C to 226°C | |
C6H5CO2CH2C≡CH | |
MFCD00150830 | |
cytidine 5'-monophosphate disodium salt, disodium cmp, cmp disodium salt, cytidine-5'-monophosphate disodium salt, disodium 5'-cmp, 5'-cmp disodium salt, disodium 5'-o-phosphonatocytidine, 5'-cytidylic acid, disodium salt, disodium cytidine 5'-monophosphate, cytidine monophosphate disodium salt | |
[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O | |
367.16 | |
160.17 |
Chemical Identifiers
6750-04-5 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only