Learn More
Propargyl alcohol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131450010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Propargyl alcohol | |
107-19-7,50-00-0 | |
Colorless to Yellow | |
114.0°C to 115.0°C | |
Authentic | |
Glass bottle | |
1.4300 to 1.4330 | |
MFCD00002912 | |
01,454 | |
0.949 | |
propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne | |
TVDSBUOJIPERQY-UHFFFAOYSA-N | |
prop-2-yn-1-ol | |
7859 | |
CHEBI:28905 | |
99% |
99% | |
-53.0°C | |
0.9490g/mL | |
34°C | |
98.5% min. (GC) | |
C3H4O | |
HC≡CCH2OH | |
1 L | |
09,394 | |
15,7923 | |
Solubility in water: miscible. Other solubilities: miscible with benzene, chloroform, ethanol, ether,, 1,2-dichloroethane, acetone, dioxane, thf,pyridine, moderately soluble in ccl4, immiscible with aliphatic hydrocarbons | |
OCC#C | |
56.06 | |
1.58 mPa.s (20°C) | |
56.06 | |
Liquid |
Chemical Identifiers
107-19-7 | |
56.06 | |
TVDSBUOJIPERQY-UHFFFAOYSA-N | |
7859 | |
prop-2-yn-1-ol |
C3H4O | |
MFCD00002912 | |
propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne | |
CHEBI:28905 | |
OCC#C |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Fatal if inhaled.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Suspected o
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 203-471-2
RTECSNumber : UK5075000
TSCA : TSCA
RUO – Research Use Only