MilliporeSigma™ PRONASE™ Protease, Streptomyces griseus, Nuclease-Free
Widely used broad-spectrum protease mixture
Supplier: MilliporeSigma™ 537088100KU
Description
- Non-specific protease liquefies mucins and digests proteins to free amino acids
- Can be used to isolate living chondrocytes
- Most active at neutral pH, but stable over wide range of pH and temperature
Specifications
Store at+2°C to+8°C | |
100kU | |
Lyophilized | |
Optimum 7.5 |